Would you like to log in again?
VWR® Critical Cover® Half-Face Shields
Supplier: Avantor
VWR® Critical Cover®
2803
47730-752CS 710.94 USD InStock
47730-752
VWR® Critical Cover® Half-Face Shields
Face Shields
Lightweight face shields feature a double-sided antifog coating
  • Low glare, zero distortion
  • Double-sided antifog coating
  • Soft polyurethane foam top for comfort
Shields also feature soft polyurethane foam at the top and a Velcro® band, allowing them to be worn comfortably over glasses for a long period of time.
Low-glare, optical grade, 7 mil thick film provides zero distortion and 100% optical clarity.
Ordering information: All shields are individually bagged and packaged 25 per dispenser box.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty