Would you like to log in again?
ValuSeal® 1500 Sealed-Edge Cleanroom Wipers, Berkshire
Supplier: Berkshire
ValuSeal®
VS1500.09098 VS1500.0909B.8
47727-366CS 1422.3 USD InStock
47727-366 47730-818
ValuSeal® 1500 Sealed-Edge Cleanroom Wipers, Berkshire
Wipes Cleanroom Wipes Dry Cleanroom Wipes
These 100% knitted polyester wipers feature double-knit construction, which makes them highly absorbent, capable of holding more liquid than many polyester knits.
  • Low particles, fibers, ions, and extractable
  • Chemically compatible with IPA, acetone, and other solvents
  • Lightweight, strong, abrasion- and tear-resistant
  • High absorbency material reduces actual cost in use compared to heavier weight products
Sealed edges ensure extremely low levels of particles, fibers, ions, and residues. For use in ISO Class 3 (FED-STD-209E Class 1/M1.5) environments.
Wipers are laundered and packaged in an ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom, minimizing the potential for contamination.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty