Would you like to log in again?
LAMP INCANDESCENT T8 LINE VOLT 100W 120V
Supplier: BULBTRONICS INC.
LAMP, INCANDESCENT^ BULB MFG PART # CEA/CEB, BULB MFG PRODUCT CODE/NAED: 1000132; 115/125V, 100W, BASE:BA15D, SHAPE:T8, OVERALL LENGTH:3 1/8", LCL: 1 3/8", LIFE:50H, VENDOR PRODUCT CODE: USCEACEB ( 0016387-USHIO), SLIDE PROJECTION LAMP
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
LAMP INCANDESCENT T8 LINE VOLT 100W 120V 47730-980 0016387 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty