Would you like to log in again?
Statshield® High Puncture Moisture Barrier Bag, Desco Industries
Supplier: Desco Industries
13760 13763 13761
47731-685EA 52.84 USD InStock
47731-685 89131-046 47731-681
Statshield® High Puncture Moisture Barrier Bag, Desco Industries
Bags Storage Bags
Statshield® Dissipative 6.5 mil Moisture Barrier Bags are designed for ESD/EMI/RFI protection.
  • Film laminate metal layered 0.0065" (6.5 mil) thick bag
  • </= 0.014 grams moisture vapor transfer rate (MVTR)
  • >30 lb. puncture strength average per FTMS 101C/4046
  • <10 nJ Energy Penetration tested per ANSI/ESD STM11.31
  • Dissipative inner and outer surfaces Rtt: < 1×10¹¹ ohms per ANSI/ESD STM11.11
  • Vacuum deposited metal layer
  • Made in the United States of America
Film laminate metal layered bags are designed for ESD/EMI/RFI protection, moisture control, high frequency and electrostatic discharge shielding. Vacuum deposited metal layer makes the bag softer, more flexible, and less likely to tear or rip during use.
Certifications: Meets JEDEC-STD-033B, ANSI/ESD S20.20, and ANSI/ESD S541 specs. Meets the requirement for static shielding per ANSI/ESD S541, ANSI/ESD S20.20, and EIA 541. Lead-free, RoHS compliant.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty