Would you like to log in again?
Statshield® Smock Jacket with Knitted Cuffs, Desco
Supplier: Desco Industries
Statshield®
73755 73772
48888-401EA 54.89 USD InStock
48888-401 47731-969
Statshield® Smock Jacket with Knitted Cuffs, Desco
Workwear
Statshield® Smock Jacket with Knitted Cuffs is a groundable static control garment system.
  • Groundable static control garment system tested per ANSI/ESD S20.20-2014 (Rtg < 3.5×10E7 ohms) requirements, per ANSI/ESD STM2.1
  • Minimum of 9% carbon conductive nylon fibers woven in chainlink design
  • Creates Faraday Cage effect around torso and arms
  • Panel to panel conductivity
  • Hip-to-cuff grounding
  • Badge holder tab
  • Two sleeve pen pockets
  • 2-year/100 launderings limited warranty
  • Made in the United States of America
Groundable Static Control Garment System allows operators and smock to be bonded or electrically connected to ground to prevent ESD events in ESD Protected Areas. Minimum of 9% carbon conductive nylon fibers woven in chainlink design provide continuous and consistent charge dissipation throughout the lifetime of the smock. Creates Faraday Cage effect around torso and arms of operator, shielding ESD charges from operators clothing from damaging ESD sensitive devices.
Hip-to-cuff grounding grounds operator with no need for a cord to be attached to the operator's wrist. Badge holder tab secures badges to avoid damage to other parts of the smock. Two sleeve pen pockets hold pens or small tools in safe and convenient location for operator.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty