Would you like to log in again?
VWR BLOCK MODULE FOR 48X0.2ML TUBES
Supplier: VWR International
Test Tube Block Module^ Holds 48x0.2mL tubes. Aluminum. Features alpha/numerical labeled rows and columns for easy vessel identification.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Unit Price Quantity
VWR BLOCK MODULE FOR 48X0.2ML TUBES 47732-408 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty