Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Analog Radio Atomic Clock
Supplier: VWR
VWR®
1077
47732-422EA 90.3 USD InStock
47732-422
VWR® Analog Radio Atomic Clock
Timers Wall Clocks
The analog radio atomic clock sets itself automatically and is accurate to 0.1 second.
  • NIST radio signal corrects time
  • Traceable
  • Bright second sweep hand is highly visible
A controlling radio signal from NIST (National Institute of Standards and Technology) in Colorado automatically adjusts the clock to reflect daylight savings time, leap year, and leap seconds (earth rotation wobble corrections).
Certifications: A serial-numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Ordering information: This clock requires one AA Battery, sold separately.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty