Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.

VWR® Digital Radio Atomic Clock

Supplier: VWR
VWR®
1076
47732-424EA 142.52 USD InStock
47732-424
VWR® Digital Radio Atomic Clock
Timers Wall Clocks
Digital clock has 92 mm (3⁵/₈") high digits for visibility from 7.6 m (25') away, and is accurate to ¹/₁₀₀₀₀ of a second.

  • Traceable®
  • Display shows AM/PM or 24-hour time, month, date, day, temperature, and humidity
  • Adjusts automatically for daylight savings, leap year, and leap seconds
  • Temperature range: 0 to 45 °C (32 to 113 °F); resolution: 0.1°
  • Relative Humidity range: 20 to 90%; resolution: 1%

Radio signal from NIST in Colorado automatically adjusts clock to precise time, daylight savings, leap year, and leap seconds (earth rotation wobble corrections).

Certifications: An individually-numbered Traceable® Certificate provided with each unit, assures accuracy from an ISO/IEC 17025:2005 (1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement.

Ordering information: Supplied Traceable® Certificate, wall mount, and batteries.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty