Would you like to log in again?
SP Bel-Art Sterileware® Sterile Mini Tongs, Bel-Art Products, a part of SP
Supplier: Bel-Art
Sterileware®
379440000
47733-014CS 68.42 USD InStock
47733-014
SP Bel-Art Sterileware® Sterile Mini Tongs, Bel-Art Products, a part of SP
Tongs
These small, lightweight tongs are ideal for sampling small quantities of material ranging from food to pharmaceuticals.
  • Lightweight
  • One-piece polystyrene plastic construction
  • Ridged jaws securely hold wet or dry samples
  • Individually packaged
  • Sterilized via gamma irradiation
Their ridged jaws securely hold wet or dry samples
Tongs are individually packaged, sterilized by gamma irradiation, lot stamped for accurate lot tracing, and shipped with a Certificate of Processing for Sterilization.
One-piece, sterile tongs are made from polystyrene plastic.
Ordering information: Individually wrapped and packed for one-time use and disposal.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty