Would you like to log in again?
Screen Prep™ Presaturated Wipes, ITW Chemtronics®
Supplier: ITW Chemtronics
Screen Prep™
CSP20
47733-170CS 268.07 USD InStock
47733-170
Screen Prep™ Presaturated Wipes, ITW Chemtronics®
Wipes Disposable Wipes and Towels Presaturated Wipes
A dry and wet wipe cleaning and static control system.
  • Dry wipes optically clean uncoated monitor screens
  • Safely remove dust, dirt, and oils
Wet wipes provide a static dissipative treatment with the convenience of a disposable applicator. Control static-generated contamination. Ideal for field service kits. Each dry wipe and wet wipe is hermetically sealed to prevent evaporation and maintain solvent effectiveness, then folded together in a twinpack. (ITW Chemtronics CSP20)
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty