Would you like to log in again?
Reaction Vessels, Cylindrical, Jacketed, Lower Drain Valves, Chemglass
Supplier: Chemglass
CG-1930-25 CG-1930-23 CG-1930-20 CG-1930-27 CG-1930-26 CG-1930-22 CG-1930-24
80071-516EA 1753.34 USD InStock
80071-516 47733-820 47733-822 47733-824 47733-826 47733-828 80071-518
Reaction Vessels, Cylindrical, Jacketed, Lower Drain Valves, Chemglass
Reaction Vessels Reaction Vessels, Cylindrical Shape
Vessels feature a cylindrical jacketed lower drain valves.
  • Similar to CG-1924, but with a choice of CG-911-A 0 to14 mm sidearm valve
  • A straight through 0 to 12 mm CG-510 valve or 0 to 20 mm CG-511 sidearm valve
All valves are designed to seal flush with the bottom of the flask avoiding the dead space normally associated with stopcock drains. This feature permits total mixing and draining of the contents of the reactor. The CG-510 is recommended for solution based reactions while the CG-911-A and CG-511 are recommended for reactions containing any solids.
Reaction vessel is constructed with a flange having a groove formed in the sealing surface of the flange. Flange is also ground to permit a greased seal with any of the lids CG-1940 through CG-1946.
Ordering information: Supplied complete with two polypropylene hose connections and two screw caps (Polypropylene hose connections with a Viton rubber seal for working temperatures up to max +110 °C) for clamp see CG-141 or CG-141-T. For O-ring see CG-147.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty