Would you like to log in again?
Cooling or Heating Coils, Stainless Steel, PFA Coated, Chemglass
Supplier: Chemglass
CG-1972-17 CG-1966-07 CG-1972-07 CG-1966-27 CG-1966-17
47734-010EA 2424.18 USD InStock
47734-010 47733-908 47734-016 47733-926 47733-918
Cooling or Heating Coils, Stainless Steel, PFA Coated, Chemglass
Heating and Cooling Coils
The cooling and heating coil is made from PFA coated stainless steel and is designed to cool or heat the contents of unjacketed cylindrical reaction vessels.
  • Greem PFA coating provides complete coverage
  • Electrically tested
  • Excellent heat transfer
Design avoids problems normally associated with heat-shrink PTFE tubing on copper tubing.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty