Would you like to log in again?
Flash Point Reference Standards, Cannon Instrument Company
Supplier: Cannon Instrument Company
9727-A20 9727-A10 9727-A25 9727-A35 9727-A15 9727-A30
47735-084EA 602.41 USD InStock
47735-084 47735-086 47735-090 47735-082 47735-092 47735-088
Flash Point Reference Standards, Cannon Instrument Company
Standards Petroleum Industry Standards
Flash point reference materials are produced in accordance with ASTM D56, Tag closed cup flash point, ASTM D92, Cleveland open cup flash point and ASTM D93, Pensky Marten closed cup flash point
Specific data including average flash point and observed standard deviation (from a statistical analysis of 40 tests in 20 diff erent laboratories) is provided with each standard.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty