Would you like to log in again?
High Viscosity Standards, Cannon Instruments
Supplier: Cannon Instrument Company
9727-E42 9727-E25.016 9727-E42 9727-E25.032 9727-E25.500 9727-E25.5 9727-E25.016 9727-E25.128 9727-E25.128 9727-E25.032
10021-364EA 2722.09 USD InStock
CA10021-364 10021-364 CA10021-362 10021-362 CA10021-360 10021-360 CA47735-156 CA10021-358 47735-156 10021-358
High Viscosity Standards, Cannon Instruments
Standards Viscometry Standards
These certified standards are useful for calibration and verification procedures.
  • For asphalt and polymer applications
Certifications: Meets the specifications of ASTM D2170 and ASTM D2171.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty