Would you like to log in again?
Viscometer Holders, Cannon
Supplier: Cannon Instrument Company
9726-N30 9726-M53 9726-M50 9726-M76 9726-N15 9726-M82 9726-N45 9726-M73 9726-M95 9726-M98 9726-M70
47735-282EA 93.4 USD InStock
47735-282 47735-292 47735-284 47735-294 47735-280 47735-290 47735-278 47735-286 47735-296 47735-288 47735-298
Viscometer Holders, Cannon
Viscometer Holders
All CANNON viscometer holders fit holes 51 mm (2" ) in diameter.
  • Self-aligning
  • Constructed of chrome-plated machined brass
  • All metal holders are provided with a handle
Plastic holders are self-aligning and hold the viscometer firmly by means of a spring clip. Rubber holders are made of neoprene rubber.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty