Would you like to log in again?
ECO Funnels, CP Lab Safety
Supplier: CP Lab Safety
EF8JUSTB2SYS EF-8-FS70-HPLC EF-3004C EF-3008 EF-4-53B-SYS EF-8-FS70 EF-4716-1P EF-8-83B EF-3004C-HPLC EF-8-JUSTRITE-B EF-4-38 EF3004CSYS EF-4-53B EF-4717-1C EF-4-38-SYS EF-30020 EF8JUSTB5SYS EF-8-JUSTRITE-B2-SYS-HPLC
33501-840EA 513.76 USD InStock
33501-840 33500-976 33500-988 33500-970 47735-370 33500-980 33500-974 47735-378 33501-844 47735-366 30246-506 47735-344 47735-346 33501-836 47735-362 33501-838 30246-504 47735-342
ECO Funnels, CP Lab Safety
Funnels Safety Funnels
ECO Funnel® is the ultimate lab safety product for collection of hazardous liquid waste. When securely attached onto an open waste container, it prevents 99.9 % of toxic fumes from being released into lab. ECO Funnel is designed to prevent fires caused by ignited vapors as well as spills of hazardous waste from open containers, which helps laboratories stay compliant with local health and safety codes.
  • Latching lid with gasket to keep fumes contained
  • Removable large particle filter catches stir bars or other items
  • Detachable tube extends into waste container to minimize chemical evaporation
The larger 8" version of ECO Funnel includes an additional safety feature to prevent over-filling; a secondary air-lock tube creates a small air gap in your waste container, which causes fluid to back up temporarily into the funnel when the container is full. After the screw cap adapter is loosened, the remaining fluid will safely flow into the container and not spill.
Ordering information: ECO Funnels can be ordered individually or as a complete system. Systems include an ECO Funnel and corresponding waste container. Some systems also include a secondary container option for maximum safety and compliance.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty