Would you like to log in again?
Amplitude™ Kappa™ Sterile LE Wipes, Contec®
Supplier: Contec
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Amplitude™ Kappa™
NWPZ0002 NWPZ0001
47735-678CS 630.73 USD InStock
47735-678 47735-676
Amplitude™ Kappa™ Sterile LE Wipes, Contec®
Wipes Cleanroom Wipes Sterile Cleanroom Wipes
Amplitude™ Kappa™ Sterile LE Wipes are made with Sontara® lyocell/polyester blend fabric. These low endotoxin wipes have excellent sorbency and are validated sterile to an SAL of 10⁻⁶.
  • Recommended for Grade A/B or ISO Class 5-6 environments
  • Peelable outer bag and linear tear inner bag allow for easy opening, even when wearing gloves
  • Lot tested and Certified low endotoxin level of <20 EU/wipe make them ideal for product contact areas
  • Suitable for use in aseptic suites and the most critical pharmaceutical cleanrooms
The packaging allows for easy opening even when wearing gloves, and the small quantities per pack minimize the possibility of waste. These wipes are ideal when increased sorbency is needed or a textured surface is required for efficient cleaning and particle removal. The wipes are also highly suitable for spill control and wiping-to-dry.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty