Would you like to log in again?
LAMP HALOGEN T3 20W 6V 100H 1.18IN
Supplier: BULBTRONICS INC.
LAMP, TUNGSTEN-HALOGEN^ TUNGSTEN-HALOGEN LAMP, 6V, 20W, BASE: PY16-1.25, SHAPE: T3, OVERALL LENGTH: 30MM, DIAMETER: 9MM, LIFE: 100H,INSTRUMENTATION LAMP
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
LAMP HALOGEN T3 20W 6V 100H 1.18IN 47738-538 0000966 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty