Would you like to log in again?
LAMP XENON ARC 150W 7.5A 20V DIA 0.79IN
Supplier: BULBTRONICS INC.
LAMP, XENON AMINCO (SLM)^ XENON LAMP AMINCO (SLM) MODEL SLM 4800, 150W, OVERALL LENGTH: 20MM, DIAMETER: 150MM, LIFE: 1200H, ALSO USED IN OTHER INSTRUMENTATION
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
LAMP XENON ARC 150W 7.5A 20V DIA 0.79IN 47742-088 0000242 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty