Would you like to log in again?
Technik™ Needle Nose Stainless Steel Pliers, Aven
Supplier: AVEN TOOLS INC.
10314
47743-090EA 60.51 USD InStock
47743-090
Technik™ Needle Nose Stainless Steel Pliers, Aven
Pliers
Pliers feature dual leaf springs and box joint construction for comfortable action.
  • Induction hardened cutting edges
  • Made of mirror polished stainless steel
  • ESD-safe grips
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty