Would you like to log in again?
Technik™ Stainless Steel Cutters, Aven Tools
Supplier: Aven
Technik™
10321 10323
47743-092EA 76.41 USD InStock
47743-092 47743-094
Technik™ Stainless Steel Cutters, Aven Tools
Cutters
These strong and versatile oval head cutters are extremely popular and used for general purpose wire cutting.
  • Box joint construction provides superior jaw alignment and durability
  • Induction hardened to 50-52 HRC
Dual leaf springs ensure opening and closing is comfortable. ESD Safe Grips provides protection against static as well as extra comfort for repetitive work. Mirror finished stainless steel (400 series) provides superior rust protection compared with standard carbon steel pliers.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty