Would you like to log in again?
Rotors for Centrifuge 5702/5702 R/5702 RH
Supplier: Eppendorf
022639471 022639421 022639421 022639048 022639501 022639048 022639471 022639501 022639304 022639064 022639404 022639064 022639480 022639404
80086-006EA 942.07 USD InStock
80086-006 CA47743-474 CA47743-482 47743-482 47743-474 CA47743-480 89207-274 47743-480 CA80086-006 80086-008 CA47743-508 47743-528 CA47743-528 47743-508
Rotors for Centrifuge 5702/5702 R/5702 RH
Centrifuges Rotors
Eppendorf rotors for centrifuge 5702 series - precision in every spin. Tailor your centrifugation with varied capacities and speeds. Accessorize with adaptable buckets for versatile sample processing. Ideal for low-speed applications, ensuring efficiency and reproducibility.
  • Precision-engineered for optimal sample separation
  • All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121 °C, 20 minutes)
  • Accessories (adaptors and buckets) further customize your centrifuge setup
Elevate your low-speed centrifugation with Eppendorf rotors designed for Centrifuge 5702 series. These rotors provide a range of capacities and speeds, offering flexibility for various applications. Accessorize with adaptable buckets, ensuring versatility in sample processing. Ideal for low-speed applications, the Centrifuge 5702 series, known for efficiency and reproducibility, is enhanced with these precision-engineered rotors. Explore the range to optimize your centrifuge for reliable performance.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty