Would you like to log in again?
Corning® ITS Universal Culture Supplement Premix, Corning
Supplier: Corning
Corning®
354351 354352 354350 354352 354351 354350
47743-624EA 510.9 USD InStock
47743-624 CACB354350 CACB354352 CACB354351 47743-626 47743-628
Corning® ITS Universal Culture Supplement Premix, Corning
Cell Culture Supplements and Additives
ITS contains insulin, human transferrin, and selenous acid, the three most universally essential components of defined culture media.
They stimulate cell proliferation of a variety of cells under serum-reduced conditions.
Tested for ability to promote proliferation of HeLa cells in serum-free medium. Filtered (0.2 µm membrane) and tested and found negative for the presence of bacteria, fungi and mycoplasma.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty