Would you like to log in again?
Corning® MITO+ Serum Extender, Corning
Supplier: Corning
Corning®
355006 355006
47743-630EA 247.6 USD InStock
47743-630 CACB355006
Corning® MITO+ Serum Extender, Corning
Cell Culture Supplements and Additives
MITO+ Serum Extender is a concentrated, fully defined formulation of hormones, growth factors (EGF and FGF), and other metabolites (insulin and steriod hormones)
It can be used to culture a variety of cells under serum-free or serum-reduced conditions.
Tested for ability to promote proliferation of BALB/c-3T3 cells in serum-reduced medium. Filtered (0.2 µm membrane) and tested and found negative for the presence of bacteria, fungi and mycoplasma.
(5ml; 5L equivalent).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty