Would you like to log in again?
Corning® Cell Recovery Solution, Corning
Supplier: Corning
Corning®
354270 354253 354253
47743-696EA 89.11 USD InStock
47743-696 CACB354253 76332-050
Corning® Cell Recovery Solution, Corning
Cell Culture Supplements and Additives
Corning® Cell Recovery Solution allows for the recovery of cells cultured on Corning® Matrigel® Basement Membrane Matrix for subsequent biochemical analysis. Corning® Cell Recovery Solution depolymerises Corning® Matrigel® Matrix gels without enzymatic digests and lengthy incubation periods at high temperatures. Cells are released without damage thereby avoiding biochemical changes during incubation and digestion of extracellular portions of cell-surface receptors and adhesion molecules.
  • Non-enzymatic proprietary solution
  • Tested for ability to depolymerise a 1 mm thick layer of gelled BD Matrigel™ matrix after one hour at 2 to 8 °C
  • Filtered and tested and found negative for bacteria, fungi, and mycoplasma
  • Stable for at least three months at 2 to 8 °C
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty