Would you like to log in again?
SP Bel-Art Microcentrifuge Tube Opener, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a part of SP
199250001
47743-758CS 1023.93 USD InStock
47743-758
SP Bel-Art Microcentrifuge Tube Opener, Bel-Art Products, a part of SP
Crimpers, Cappers and Decappers
This convenient tool for opening microcentrifuge tubes is double-ended to work on all tube sizes.
  • Double-ended design
  • Open milli- and microliter-sized tubes
  • Convenient design and handling
One end opens standard 1.5, 2.0, and 2.2 mL tubes while the other end has a forked blade for opening smaller sized tubes — 250, 400, and 550µL.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty