Would you like to log in again?
PLATE DEEP WELL 500UL PK5
Supplier: Corning
Deep Well Plate^ 85.4x127.7mm to near corners, individual well 6.5x29.0mm, manufactured from 99.9% pure virgin polypropylene, guaranteed RNase, DNase and pyrogen free
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
PLATE DEEP WELL 500UL PK5 47743-828 P-DW-500-C Case of 50 Retrieving
Pack of 5 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty