Would you like to log in again?
SatPax® ValuSeal® 1500 Presaturated Cleanroom Wipers, Berkshire
Supplier: Berkshire
SatPax®
SPXV15001.12
47744-010CS 813.74 USD InStock
47744-010
SatPax® ValuSeal® 1500 Presaturated Cleanroom Wipers, Berkshire
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
Laser-cut, sealed-edge, 100% double-knit polyester wipers saturated with semiconductor-grade isopropyl alcohol and semiconductor-grade reverse osmosis/deionized water.
  • SatPax® HA combines ValuSeal® HA ultrasonically sealed edge cleanroom laundered wipers composed of 100% continuous filament
  • Polyester knit fabric with a high saturation level of 70% IPA and 30% DI water
  • 100% continuous filament polyester knit
  • Laundered and packaged in Berkshire’s ISO Class 4 cleanroom
  • Re-sealable solvent resistant packaging
  • Wipers in J-folded configuration for single withdrawal
Note: Specifications refer to wiper material prior to saturation.
Combination of solvent and wiper improves the cleaning process and resolves safety issues associated with storage, mixing, and dispensing of solvents.
Ordering information: Laundered and packaged in an ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom. Packaged in convenient, resealable packs.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty