Would you like to log in again?
Counter Tops with Flat Top, Stainless Steel, VCTC Series, Advance Tobaco
Supplier: Advance Tabco
VCTC-245 VCTC-248
47744-714EA 5893.89 USD InStock
47744-714 47744-708
Counter Tops with Flat Top, Stainless Steel, VCTC Series, Advance Tobaco
Furniture Tables and Workbenches
The VCTC series is furnished with 1.5×1" square die embossed No-Drip countertop edge with a 0.5" return on four sides.
  • 16 gauge stainless steel type 304
  • All TIG welded
  • Exposed weld areas polished to match adjacent surfaces
  • Entire top mechanically polished to a satin finish
  • Countertop edge polished to a mirror finish
  • Top is sound deadened
  • NSF
Roll formed embossed galvanized hat channels are secured to top by means of structural adhesive and weld studs. Engineered and readily adaptable for installation on standard kitchen cabinets. No backsplash.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty