Would you like to log in again?
Staticide® Restorer & Cleaner, Anti-Static Floor Cleaner, ACL
Supplier: ACL Staticide
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Staticide®
4100-5 4100-1
47744-782EA 317.46 USD InStock
47744-782 47744-780
Staticide® Restorer & Cleaner, Anti-Static Floor Cleaner, ACL
Cleaning Agents Detergents and Surface Cleansers
ACL Staticide Restorer Cleaner is a specialized anti-static detergent for ESD floor finishes. The cleaner renews static decay properties, surface resistivity, and the durability of ACL floor finishes. Special conductive additives clean while simultaneously breaking down electrical discharge and returning floor finishes to safe specifications.
  • Removes black heel marks
  • Lengthens maintenance cycle
  • Revitalizes gloss
  • Dilution ratio 1:10 (1 part cleaner to 10 parts water)
  • Covers approximately 1000 - 1500 square feet per diluted gallon
No harmful chemicals like ammonia, phosphates, alkali, nonylphenol (NP), or nonylphenol ethoxylate (NPE) are used in the formulation. This special detergent can be applied with a mop or in a standard burnishing machine for easy application.
Ideal for the following applications:
• Electronic manufacturing facilities
• Maintaining ESD Floor Finishes
• Maintaining ESD Floor Tiles
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty