Would you like to log in again?
ep Dualfilter T.I.P.S.® Racks, Filter Pipette Tips
Supplier: Eppendorf
ep Dualfilter T.I.P.S.®
0030078810 022491288 022491288 0030078551 0030078845 0030078829 0030078594 022491211 0030078624 0030078586 0030078535 022491237 022491202 0030078519 022491245 0030078616 0030078594 0030078578 022491229 022491270 022491253 0030078586 0030078543 0030078551 0030078500 0030078527 0030078829 0030078616 0030078560
47745-088CS 233.26 USD InStock
47745-088 76018-998 47747-994 CA47745-092 CA47745-090 CA89014-311 47745-084 CA47745-096 47745-086 89126-924 CA47745-094 CA76423-882 CA89014-308 CA47747-994 CA47745-088 CA47745-086 CA76018-994 47745-092 CA89126-924 47745-094 47745-096 CA47745-084 89014-309 75845-010 47745-090 76423-882 76018-994 76018-992 89132-036
ep Dualfilter T.I.P.S.® Racks, Filter Pipette Tips
Pipette Tips
Eppendorf ep Dualfilter T.I.P.S.® Racks offer 'PCR clean and Sterile' grade filter pipette tips, providing dual-layer protection against contamination for critical lab applications, ensuring precision, reliability, and hygiene in high-stakes environments.
  • Two filter layers provide dual protection against aerosols and biomolecules to guard both pipette and sample
  • Free of PCR inhibiting additives provide for reliable downstream results
  • Available in PCR clean/Sterile (sterile and pyrogen-free)
  • Continuous inspection of each production lot by an independent laboratory guarantees high purity and quality
  • Modern slim rack design with reclosable lid
  • Improved sustainability through 20% lower polypropylene consumption per rack
  • ACT® labeled by non-profit My Green Lab® to help make an informed choice regarding sustainability
Take advantage of an additional barrier in filter pipette tips:
ep Dualfilter T.I.P.S.® from Eppendorf are filter pipette tips with a two-phase filter for contamination protection. The two filter layers, made of flexible, hydrophobic material, fit perfectly in the tip cone and retain practically 100% of all aerosols and biomolecules. This filtering effect is achieved using various, well-defined pore sizes. The layer facing the sample provides protection from drops, splashes and aerosols. The layer facing the pipette tip cone functions as a second barrier from contamination, and reliably binds biomolecules. A defined air passage rate ensures full sample recovery and quick pipetting.
EPA/HEPA Standard - Filter efficiency tested and certified for ep Dualfilter T.I.P.S.®:
ep Dualfilter T.I.P.S.® and ep Dualfilter T.I.P.S.® SealMax filter pipette tips have even been tested for filtration performance under the standard EN 29463 (Standard of filtration performance testing of filters), yielding a minimum particle collection efficiency of 99.5% with test aerosol particles sizes of 0.05 to 0.5 µm. They are ideal for applications in cleanrooms and in the pharmaceutical industry.
ep Dualfilter T.I.P.S.® in the purity grade PCR clean/Sterile are sterilized by electron beams, certified pyrogen-free and PCR clean (free from human DNA, DNase, RNase, and PCR-inhibiting substances). ep Dualfilter T.I.P.S.® Forensic DNA Grade are free of detectable Human DNA, DNase, RNase, and PCR inhibitors.
Certifications: Lot-specific certficates available for the purity grades PCR clean/Sterile, Forensic DNA Grade can be downloaded from the Eppendorf website.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty