Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

epT.I.P.S.® Reloads, Pipette Tips

Supplier: Eppendorf
epT.I.P.S.®
022491521 022491768 022491512 022491563 022491547 022491776 022494006 022491512 022491539 022494004 022491768 022491571 022491571 022491717 022491504 022494006 022491709 022491547 022494004 022491709 022491725 022491717 022491555 022491539 022491741 022491741 022491733 022491750 022491521 022491750 022491504 022491725 022491563 022491733 022491555 022491776
47745-144CS 95.51 USD InStock
47745-144 CA47745-132 CA47745-154 47745-146 CA47745-152 CA47745-136 47745-148 CA47745-158 CA47745-134 CA47745-156 89126-926 47745-160 89126-928 47745-162 47745-140 CA47745-150 47745-142 47745-132 CA47745-144 47745-154 47745-134 47745-156 CA47745-142 CA89126-928 CA47745-148 47745-158 47745-136 47745-138 CA47745-146 CA89126-926 CA47745-140 47745-150 CA47745-162 CA47745-160 47745-152 CA47745-138
epT.I.P.S.® Reloads, Pipette Tips
Pipette Tips
Eppendorf epT.I.P.S.® Reloads offer high-precision pipette tips in 'Eppendorf Quality' and 'PCR clean' purity grades. Eco-friendly, cost-effective, and designed for seamless compatibility with Eppendorf Box 2.0, they ensure accuracy and sustainability in labs.

  • Reduced waste when compared with disposable racks
  • Refill trays can be autoclaved up to 121 °C, in packaging and without foil, for subsequent use
  • Depending on tip size, the refill system is packaged as either dual-sided or in stack form
  • Available in purity grades of Eppendorf Quality and PCR clean
  • Color-coded trays for simple volume and pipette identification
  • ACT® labeled by non-profit My Green Lab® to help make an informed choice regarding sustainability

The perfect fit for Eppendorf pipettes: epT.I.P.S.® ensure a reliable pipetting system:
Make everyday work easier and more enjoyable with the epT.I.P.S.® (Eppendorf "Totally Integrated Pipetting System") pipette tips. These pipette tips are manufactured to extremely high quality and purity standards and are designed to cover all types of specific sampling needs. Offering reduced-force tip attachment and ejection with complete sealing, epT.I.P.S.® work in perfect combination with Eppendorf pipettes and form a fully integrated pipetting system that delivers maximum precision, accuracy and reproducibility. Eppendorf pipette tips with ideal wetting properties and optimum transparency are manufactured exclusively from high-purity plastic materials. The universal nose cone design allows Eppendorf tips to also be used with pipettes from other manufacturers.

How to recognize an original Eppendorf pipette tip?
All Eppendorf tips feature a 'ring' of drop-shaped relief elements on the edge of the tip crown.

Extended length for reliable sampling: epT.I.P.S.® L pipette tips:
The elongated pipette tips (epT.I.P.S.® L) enable reliable sample reach while pipetting from and into conical tubes, high reagent bottles, narrow deep vessels, cell culture flasks, or deepwell plates. The long and slim design of these elongated pipette tips allows free access to the sample with reduced risk of touching the sides of tubes or wells. Cross-contamination during pipetting can be reduced to a minimum. epT.I.P.S.® L are available in volumes of: 0.5 to 20 µl, 50 to 1250 µl, 0.2 to 5 ml, and 0.5 to 10 ml.

Certifications: Lot-specific certificates for the purity grade PCR clean can be downloaded from the Eppendorf website.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty