Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

epT.I.P.S.® Bulk - Pipette Tips

Supplier: Eppendorf
epT.I.P.S.®
022492071 022492021 0030000650 022492012 022492071 022492098 022492080 022492004 022492021 022492004 022494018 022492101 022492080 022492039 022492047 022492063 022492039 022494018 022492055 022492012 0030000650 022492063 022492098 022492055 022492101 022492047
47745-166CS 103.55 USD InStock
47745-166 CA47745-176 47745-168 CA47745-174 CA75845-008 CA47745-178 47745-180 CA89126-938 47747-992 CA47745-172 47745-164 CA47745-170 47747-986 47745-176 CA47745-166 47745-178 CA47745-164 CA47745-168 CA47747-992 89126-938 CA47745-180 47745-170 75845-008 47745-172 47745-174 CA47747-986
epT.I.P.S.® Bulk - Pipette Tips
Pipette Tips
Eppendorf epT.I.P.S.® Bulk packs offer a practical, high-volume solution with 'Eppendorf Quality' pipette tips, ensuring precise and reliable measurements for various lab applications, packaged economically for high-throughput settings.

  • Original, high-quality Eppendorf pipette tips packaged in resealable bags
  • Available in sizes from 10 µl to 10 ml in Eppendorf Quality purity grade
  • epT.I.P.S.® Standard 200, 300 and 1000 µl tips are also available color-coded yellow and blue
  • epT.I.P.S.® Standard pipette tips can be autoclaved at 121 °C in 20 min
  • Fine retracted or extended tips simplify work with small volumes
  • ACT® labeled by non-profit My Green Lab® to help make an informed choice regarding sustainability

The perfect fit for Eppendorf pipettes: epT.I.P.S.® ensure a reliable pipetting system:
Make everyday work easier and more enjoyable with the epT.I.P.S.® (Eppendorf 'Totally integrated pipetting system') pipette tips. These pipette tips are manufactured to extremely high quality and purity standards and are designed to cover all types of specific sampling needs. Offering reduced-force tip attachment and ejection with complete sealing, epT.I.P.S.® work in perfect combination with Eppendorf pipettes and form a fully integrated pipetting system that delivers maximum precision, accuracy, and reproducibility. Eppendorf pipette tips with ideal wetting properties and optimum transparency are manufactured exclusively from high-purity plastic materials. The universal nose cone design allows Eppendorf tips to also be used with pipettes from other manufacturers.

How to recognize an original Eppendorf pipette tip?
All Eppendorf tips feature a 'ring' of drop-shaped relief elements on the edge of the tip crown.

Extended length for reliable sampling: epT.I.P.S.® L pipette tips:
The elongated pipette tips (epT.I.P.S.® L) enable reliable sample reach while pipetting from and into conical tubes, high reagent bottles, narrow deep vessels, cell culture flasks, or deepwell plates. The long and slim design of these elongated pipette tips allows free access to the sample with reduced risk of touching the sides of tubes or wells. Cross-contamination during pipetting can be reduced to a minimum. epT.I.P.S.® L are available in volumes of: 0.5 to 20 µl, 50 to 1250 µl, 0.2 to 5 ml, and 0.5 to 10 ml.

Certifications: General quality and ISO certificates available.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty