Would you like to log in again?
epT.I.P.S.® Racks - Pipette Tips
Supplier: Eppendorf
epT.I.P.S.®
0030075293 0030075307 022491113 022491083 0030071620 022491989 0030071581 0030071620 0030075293 0030075277 022491881 022491091 0030071573 022491105 022491067 022491121 0030071646 0030071638 0030071646 0030071590 0030075269 0030075307 0030075226 0030075285 0030071557 0030071581 022491164 0030075277 0030071654 0030075242 0030075250 0030071565 0030075234 0030071573 022491997 0030071565 0030071662
47745-080CS 141.7 USD InStock
CA47745-198 CA47745-078 CA47751-660 CA47745-076 47745-080 CA76423-878 CA47751-668 47745-082 47751-659 CA76423-876 47747-990 47751-657 CA47745-074 CA47751-664 CA89126-934 CA47745-072 CA76423-880 47747-988 47745-078 47745-198 47751-661 47745-216 CA47745-080 CA47751-658 47751-669 47745-072 47745-074 47751-667 89126-934 76423-876 47745-076 CA47745-082 47751-665 76423-878 CA47747-988 76423-880 CA47745-216
epT.I.P.S.® Racks - Pipette Tips
Pipette Tips
Eppendorf epT.I.P.S.® Racks feature high-precision pipette tips in 'Eppendorf Quality', 'PCR clean', 'Biopur®', and 'Sterile' purity grades. All tips are supplied in disposible racks designed for optimal organization and user convenience in demanding laboratory environments.
  • Long, narrow tips reach the base of narrow and high vessels without touching the top of the vessel edge
  • Modern slim rack design with reclosable lid
  • Improved sustainability through 35% lower polypropylene consumption per rack
  • ACT® labeled by non-profit My Green Lab® to help make an informed choice regarding sustainability
The perfect fit for Eppendorf pipettes: epT.I.P.S.® ensure a reliable pipetting system:
Make everyday work easier and more enjoyable with the epT.I.P.S.® (Eppendorf 'Totally integrated pipetting system') pipette tips. These pipette tips are manufactured to extremely high quality and purity standards and are designed to cover all types of specific sampling needs. Offering reduced-force tip attachment and ejection with complete sealing, epT.I.P.S.® work in perfect combination with Eppendorf pipettes and form a fully integrated pipetting system that delivers maximum precision, accuracy, and reproducibility. Eppendorf pipette tips with ideal wetting properties and optimum transparency are manufactured exclusively from high-purity plastic materials. The universal nose cone design allows Eppendorf tips to also be used with pipettes from other manufacturers.
How to recognize an original Eppendorf pipette tip?
All Eppendorf tips feature a 'ring' of drop-shaped relief elements on the edge of the tip crown.
Extended length for reliable sampling: epT.I.P.S.® L pipette tips:
The elongated pipette tips (epT.I.P.S.® L) enable reliable sample reach while pipetting from and into conical tubes, high reagent bottles, narrow deep vessels, cell culture flasks, or deepwell plates. The long and slim design of these elongated pipette tips allows free access to the sample with reduced risk of touching the sides of tubes or wells. Cross-contamination during pipetting can be reduced to a minimum. epT.I.P.S.® L are available in volumes of: 0.5 to 20 µl, 50 to 1250 µl, 0.2 to 5 ml, and 0.5 to 10 ml.
Certifications: Lot-specific certficates available for the purity grades PCR clean, Sterile, and Biopur® can be downloaded from the Eppendorf website.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty