Would you like to log in again?
Axygen® Robotic Pipette Tips, Corning
Supplier: Corning
Axygen®
ZT-100-R TWP-200-R-S PK-50-R LT-384-R PK-200-R
80078-802CS 788.57 USD InStock
80078-802 47734-686 80082-910 47745-460 47745-472
Axygen® Robotic Pipette Tips, Corning
Pipette Tips
Axygen® brand automation tips are engineered to the exact specifications of the original equipment manufacturers’ tips, ensuring complete compatibility and a consistent fit.
  • Engineered to the exact specifications to ensure compatibility and consistent fit
  • Certified DNase-/RNase-free and nonpyrogenic for complete protection against contamination
  • Maxymum Recovery® versions also available
  • Filtered version available in most tip styles to prevent aerosol and liquid contamination
Axygen® offers a comprehensive line of tips for Beckman Coulter®, Dynamic Devices, Hamiltion Starline, and ProGroup Wellpro automation systems. These tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality. Available in nonsterile, sterile, non-filtered, filtered, wide-bore, fine point, and extended length formats. Clear polypropylene tips are available in 96-well and 384-well formats. Also available in Maxymum Recovery® to further improve accuracy and precision while reducing sample loss.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty