Would you like to log in again?
Pleated (Fluted) Filter Paper, Grade 515, Ahlstrom
Supplier: Ahlstrom
5150-1850 5150-2400 5150-1250
38000-200CS 3403.27 USD InStock
38000-200 47745-846 47745-848
Pleated (Fluted) Filter Paper, Grade 515, Ahlstrom
Filters Filter Paper Qualitative Filter Paper
Ahlstrom Grade 515 is the pre-pleated version of Grade 615.
Pleating improves the effective surface area which increases flow rate and loading capacity, thus decreasing filtration time.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty