Would you like to log in again?
Stainless Steel Service Sinks, Advance Tabco®
Supplier: Advance Tabco
4-OP-18 8-OP-16
47747-438EA 4385.11 USD InStock
47747-438 47747-436
Stainless Steel Service Sinks, Advance Tabco®
Furniture Casework
This conventional sink is fabricated from heavy gauge stainless steel and has a deep drawn bowl.
Economy sink is a welded bowl style. Both come with 3.8 cm (11/2") IPS basket-type drain. Legs are constructed of stainless steel tubing with 2.5 cm (1") adjustable feet. Both sinks are TIG welded for strength.
Ordering information: Faucet should be ordered seperately.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty