Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

SP Bel-Art Precisionware® Bottles, Narrow Mouth, Amber, HDPE, Bel-Art Products, a part of SP

Supplier: Bel-Art

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

SCIENCEWARE®
10624-9015 10624-9007 10624-9016 10624-9008
47748-104CS 693.47 USD InStock
47748-104 47748-100 47748-102 47748-098
SP Bel-Art Precisionware® Bottles, Narrow Mouth, Amber, HDPE, Bel-Art Products, a part of SP
Bottles Plastic Bottles
FDA compliant laboratory/pharmaceutical grade, amber colored bottles are perfect for the storage and sampling of light sensitive materials.

  • Rigid HDPE construction
  • Polypropylene closures
  • Leakproof design
  • Amber coloring provides protection for light-sensitive materials

Made of rigid high-density polyethylene, they have matched polypropylene closures with a leakproof design. They are ideal for packaging and storage of liquids and powders. Bottles 250 ml (8.4 oz.) and larger feature a recycling code molded into the base.

Certifications: Bottles comply with U.S. Pharma 23 requirements for light transmission.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty