Would you like to log in again?
Nitrilite® 93-401, Silky Ultra-Clean, Gloves, Ansell
Supplier: Ansell
Nitrilite™
9340111L 9340111XXL 365760 365772 9340111M 365759 365761 365758 9340111XL 9340111S 365757
80093-140CS 597.94 USD InStock
CA80093-140 CA47748-238 80093-140 47748-236 CA47748-234 CA47748-236 47748-238 47748-232 47748-234 CA47748-232 47748-230
Nitrilite® 93-401, Silky Ultra-Clean, Gloves, Ansell
Gloves
These ambidextrous gloves have the superior strength of nitrile yet are thinner than comparable vinyl or latex gloves for a lighter, more comfortable glove with improved dexterity and tactility.
  • Beaded cuffs
  • Powder-free
  • Strong, yet light and flexible
These gloves significantly reduce particlate and extractable levels.
Ideal applications include electronics assembly, handling sensitive parts and materials, pharmaceutical manufacturing and product protection in handling cosmetics, packaging, optical goods, microelectronic and micro-mechanic components, as well as protection from blood and other bodily fluids.
They are powder-free with no plasicizers, silicone or pigments.
Ordering information: Gloves are packaged in an ISO Class 5 (FED-STD-209E Class 100/M3.5) environment. Cleanroom-compatible packaging contains 50 gloves per inner poly bag and 2 inner bags per outer poly bag. Lot number is printed on outer poly bag in IPA-resistant ink.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty