Would you like to log in again?
Hardened Ashless Quantitative Paper, Grade 55, Ahlstrom
Supplier: Ahlstrom
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
0550-2700 0550-1500 0550-4000 0550-1100 0550-1850 0550-3200 0550-0470 0550-5000 0550-0700 0550-2400 0550-0900 0550-1250 0550-0425 0550-0550
28334-145PK 87.87 USD InStock
28334-145 28334-154 47749-460 47749-450 47749-454 47749-462 47749-452 47749-458 47749-456 47749-448 47745-736 38008-027 47745-738 38008-129
Hardened Ashless Quantitative Paper, Grade 55, Ahlstrom
Filters Filter Paper Quantitative Filter Paper
Ahlstrom Grade 55 is a fast filtering hardened ashless (Ash 0.006%) filter paper suitable for quantitative gravimetric analysis of coarse particles and gelatinous precipitates in analytical techniques requiring increased wet strength and handling capacity.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty