Would you like to log in again?
Glass Microfiber Filter Paper, Grade 121, Ahlstrom
Supplier: Ahlstrom
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
1210-0700 1210-0425 1210-0210 1210-0370 1210-0900 1210-0470 1210-1250 1210-1100 1210-0550 1210-0240 1210-1500 1210-0250
47745-794PK 235.39 USD InStock
47745-794 47749-534 47745-792 47749-532 38010-436 47749-538 47749-536 47745-790 47749-542 38010-429 38010-418 47749-540
Glass Microfiber Filter Paper, Grade 121, Ahlstrom
Filters Glass Fiber Filters
Grade 121 is a very thick binder-free microfiber glass filter paper that offers finer particle retention than Ahlstrom 111 with higher wet strength and loading capacity.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty