Would you like to log in again?
Glass Microfiber Filter Paper Grade 151, Ahlstrom
Supplier: Ahlstrom
1510-0470 1510-0550 1510-0425 1510-0210 1510-0240 1510-0250 1510-1500 1510-0700 1510-0900 1510-1250 1510-1100 1510-1420 1510-0370
28496-178PK 404.94 USD InStock
28496-178 47749-582 47749-580 47749-586 47749-584 47749-578 28496-180 47749-588 28496-174 28496-176 38010-838 28496-182 28496-184
Glass Microfiber Filter Paper Grade 151, Ahlstrom
Filters Glass Fiber Filters
Ahlstrom Grade 151 is a binder-free microfiber glass filter designed to retain extremely fine particles.
It is recommended for use in the Toxicity Characteristic Leaching Procedure (TCLP) as described in EPA Method 1311.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty