Would you like to log in again?
Qualitative Filter Paper, Grade 601, Ahlstrom
Supplier: Ahlstrom
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
6010-1500 6010-2700 6010-3850 6010-1100 6010-3200 6010-0300 6010-1250 6010-0750 6010-0250 6010-0900 6010-2500 6010-4000 6010-1850 6010-0550 6010-0700 6010-0470 6010-2400 6010-0425 6010-5000
38000-025BX 20.29 USD InStock
38000-025 47745-810 38000-229 47749-670 47749-662 47749-660 47749-666 47749-656 47749-664 47745-808 47749-668 47749-658 38000-160 38000-182 47745-814 47745-812 28342-016 47745-818 47745-816
Qualitative Filter Paper, Grade 601, Ahlstrom
Filters Filter Paper Qualitative Filter Paper
Ahlstrom Grade 601 is used in a wide range of laboratory applications such as clarifying liquids, rapid filtration of coarser precipitates, soil analysis, seed testing and more.
This low ash grade contains more than 95% super refined alpha-cellulose and linter fibers.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty