Would you like to log in again?
Qualitative Filter Paper, Grade 631, Ahlstrom
Supplier: Ahlstrom
6310-0550 6310-1500 6310-1250 6310-1100 6310-3200 6310-0425 6310-2500 6310-0900 6310-2400 6310-1850 6310-2700
47749-688PK 12.85 USD InStock
47749-688 47749-700 47749-686 47749-698 47749-704 47749-702 47749-708 47749-692 47749-706 47749-696 47749-694
Qualitative Filter Paper, Grade 631, Ahlstrom
Filters Filter Paper Qualitative Filter Paper
Ahlstrom Grade 631 is used for filtration of coarse particles and some gelatinous precipitates, air monitoring when high flow rate is needed, and in routine cleanup analysis of insoluble liquids.
This low ash grade contains more than 95% super refined alpha-cellulose and linter fibers.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty