Would you like to log in again?
Wet-Strengthened Qualitative Filter Papers, Grade 962
Supplier: Ahlstrom
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
9620-1250 9620-1500 9620-0900 9620-0550 9620-5000 9620-4000 9620-2400 9620-1850 9620-0425 9620-2700 9620-1100 9620-3200 9620-0700
47749-790PK 16.2 USD InStock
47749-790 47749-794 47749-792 47749-798 47749-810 47749-796 47749-802 47749-814 47749-800 47749-812 47749-806 47749-804 47749-808
Wet-Strengthened Qualitative Filter Papers, Grade 962
Filters Filter Paper Qualitative Filter Paper
Ahlstrom Grade 962 is a fast filtering grade with a creped surface and high loading capacity.
It is a thinner version of Grade 961.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty