Would you like to log in again?
SP Bel-Art Heavy Duty Faucets, Bel-Art Products, a part of SP
Supplier: Bel-Art
SCIENCEWARE®
H30857-0000 F30851-0000 F30852-0000
47750-018EA 79.67 USD InStock
47750-018 47750-014 47750-016
SP Bel-Art Heavy Duty Faucets, Bel-Art Products, a part of SP
Faucets and Sinks Faucets
A needle-type, Teflon®O-Ring seal inside this polyethylene faucet prevents back pressure liquid from escaping through the operating knob.
  • Polyethylene construction
  • Needle-type seal
  • Secure against back pressure liquid escaping
The outlet accepts 16 mm I.D. (5/8") tubing.
Faucet 47750-016 fits standard steel drums. Faucet 47750-018 has a male threaded outlet that accommodates garden hose coupling standard 19 mm – 4.53 threads per cm (3/4" – 11.5 threads per inch).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty