Would you like to log in again?
SP Bel-Art Three-Way Fluoroplastic Stopcocks, Bel-Art Products, a part of SP
Supplier: Bel-Art
F30888-0010 F30888-0008 F30888-0006
47750-034EA 141.11 USD InStock
47750-034 47750-036 47750-038
SP Bel-Art Three-Way Fluoroplastic Stopcocks, Bel-Art Products, a part of SP
Stopcocks
The three-way configuration of these stopcocks permits connection of any two or all three ports.
  • PVDF construction
  • Chemically resistant
  • Autoclavable
Barbed tubulations accept rubber or plastic tubing. They are made of PVDF (polyvinylidene fluoride), a fluoroplastic resistant to most chemicals that withstands temperatures up to 204ºC (400ºF). Maximum pressure rating 34.3 KPa (5 psi). Steam autoclavable.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty