Would you like to log in again?
SP Bel-Art Stopcocks, Bel-Art Products, a part of SP
Supplier: Bel-Art
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
SCIENCEWARE®
F30891-0000 F30890-0000 F30894-0000 F30895-0000
47750-040EA 64.42 USD InStock
47750-040 47750-042 47750-044 47750-046
SP Bel-Art Stopcocks, Bel-Art Products, a part of SP
Stopcocks
With a removable, leak-proof plug made of chemical-resistant, non-stick Teflon® PTFE, these 2- and 3-way polypropylene plastic stopcocks have barbed tubulations that accep 6.4 to 9.5 mm (¼ to ⅜”") I.D.tubing.
  • Leakproof plug made of chemically resistant PTFE
  • Polypropylene plastic stopcock construction
  • Plugs are non-sticking
  • Autoclavable
Three-way models allow connection of any two or all three arms. Pressure rating is 240KPa (35psi). Steam autoclavable at 121°C (250°F).
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty