Would you like to log in again?
SP Bel-Art Spinbar® Jumbo Magnetic Retriever, Bel-Art Products, a part of SP
Supplier: Bel-Art
SCIENCEWARE®
F37781-0000
47750-234EA 372.01 USD InStock
47750-234
SP Bel-Art Spinbar® Jumbo Magnetic Retriever, Bel-Art Products, a part of SP
Stirrers Magnetic Stirring Bars
This flexible PTFE magnetic retriever will remove the largest magnetic stirring bars from deep vessels.
  • PTFE construction
  • Easily remove magnetic stirring bars
  • Easy-grip handle
Retriever has an easy grip “T” handle, with an overall length of 65 cm (26"). This includes the powerful Alnico magnetic end, which is 10.2 x 1 cm diameter (4 x 3/8").
Ordering information: Longer, custom lengths are available.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty