Would you like to log in again?
SP Bel-Art Desiccator Cabinet, Acrylic, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a part of SP
F42066-0001
47750-258EA 831.73 USD InStock
47750-258
SP Bel-Art Desiccator Cabinet, Acrylic, Bel-Art Products, a part of SP
Desiccators Desiccator Cabinets
These durable cabinets are made from 6.35 mm thick acrylic.
  • Durable acrylic construction
  • Dry, dust-free storage
  • For use in freezers, refrigerators, or on the benchtop
  • Supplied with perforated trays
  • Ensured proper circulation within the cabinet
The gasketed, airtight doors are made from 9.5 mm thick acrylic with full-length hinges and stainless steel twist locking latches. Available in clear or dark bronze acrylic.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty